Methyl 6-chloro-1H-indazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A326337-100MG A326337-1G A326337-250MG A326337-5G
77015-344EA 21.22 USD
77015-344 77015-346 77015-348 77015-350
Methyl 6-chloro-1H-indazole-3-carboxylate 97%
Methyl 6-chloro-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
MDL Number: MFCD07371602
CAS Number: 717134-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR