2-Azabicyclo[2.2.2]octan-3-one 97%
Supplier: Ambeed
|
Warning
|
A325826-100MG
A325826-1G
A325826-250MG
A325826-5G
77032-802EA
77.45
USD
77032-802
77032-804
77032-806
77032-808
2-Azabicyclo[2.2.2]octan-3-one 97%
2-Azabicyclo[2.2.2]octan-3-one
Precaución: for Research Use Only
Formula:
C₇H₁₁NO |
MDL Number:
MFCD16547516 CAS Number: 3306-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...