3,5-Diiodopyridin-4-ol 96%
Supplier: Ambeed
|
Warning
|
A325741-1G
A325741-250MG
A325741-5G
A325741-25G
77015-338EA
18.04
USD
77015-338
77015-340
77015-342
77612-878
3,5-Diiodopyridin-4-ol 96%
3,5-Diiodopyridin-4-ol
Precaución: for Research Use Only
Formula:
C₅H₃I₂NO MW: 346.89 g/mol |
MDL Number:
MFCD00023554 CAS Number: 7153-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...