N,1-Di-Boc-L-histidine dicyclohexylammonium salt 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-His(Boc)-OH.DCHA, N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, Boc-His(Boc)-OH·DCHA
A325717-100G
A325717-10G
A325717-1G
A325717-25G
A325717-5G
A325717-500G
77002-928EA
137
USD
77002-928
77002-930
77002-932
77002-934
77002-936
77881-911
N,1-Di-Boc-L-histidine dicyclohexylammonium salt 97%
N,1-Di-Boc-L-histidine dicyclohexylammonium salt
Precaución: for Research Use Only
Formula:
C₁₆H₂₅N₃O₆·C₁₂H₂₃N MW: 536.714 g/mol |
MDL Number:
MFCD06804675 CAS Number: 31687-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...