3-Chloro-5-iodophenol 98%
Supplier: Ambeed
|
Warning
|
A325639-10G
A325639-1G
A325639-25G
A325639-250MG
77015-334EA
231
USD
77015-334
77015-336
77571-928
77890-649
3-Chloro-5-iodophenol 98%
3-Chloro-5-iodophenol
Precaución: for Research Use Only
Formula:
C₆H₄ClIO |
MDL Number:
MFCD12405417 CAS Number: 861347-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...