5-Bromothiophene-3-carboxamide 96%

Supplier: Ambeed
Warning

A325615-100MG A325615-10G A325615-250MG A325615-1G
76989-202EA 29 USD
76989-202 76989-204 76989-206 77612-874
5-Bromothiophene-3-carboxamide 96%
5-Bromothiophene-3-carboxamide

Precaución: for Research Use Only

Formula: C₅H₄BrNOS
MDL Number: MFCD11644062
CAS Number: 189329-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR