1-Methylpyrrolidine-2-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A325513-100MG A325513-250MG
77032-798EA 19.1 USD
77032-798 77032-800
1-Methylpyrrolidine-2-carboxylic acid hydrochloride 97%
1-Methylpyrrolidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD18071558
CAS Number: 30727-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR