(R)-2-Benzyl-4-(tert-butoxy)-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A325491-100MG A325491-1G A325491-250MG
76989-196EA 116.7 USD
76989-196 76989-198 76989-200
(R)-2-Benzyl-4-(tert-butoxy)-4-oxobutanoic acid 97%
(R)-2-Benzyl-4-(tert-butoxy)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₄
MW: 264.31 g/mol
MDL Number: MFCD12755810
CAS Number: 122225-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR