2-Bromo-5-fluoro-4-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluoro-4-methylnitrobenzene, 1-Bromo-4-fluoro-2-methyl-5-nitrobenzene

A325482-100MG A325482-1G A325482-250MG A325482-25G A325482-5G
77015-324EA 5 USD
77015-324 77015-326 77015-328 77015-330 77015-332
2-Bromo-5-fluoro-4-nitrotoluene 97%
2-Bromo-5-fluoro-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD11110550
CAS Number: 170098-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR