4-Chloro-2,7-dimethylquinazoline 97%

Supplier: Ambeed
Warning

A325336-100MG A325336-1G A325336-250MG
77015-314EA 78.51 USD
77015-314 77015-316 77015-318
4-Chloro-2,7-dimethylquinazoline 97%
4-Chloro-2,7-dimethylquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂
MW: 192.64 g/mol
MDL Number: MFCD22397495
CAS Number: 1444351-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR