1-Ethyl-1H-pyrazole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A325316-250MG A325316-1G A325316-5G
77032-796EA 12 USD
77032-796 77872-467 77872-468
1-Ethyl-1H-pyrazole-3-carbaldehyde 98%
1-Ethyl-1H-pyrazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD03419802
CAS Number: 942319-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR