4-Bromobenzo[d]isoxazole 97%

Supplier: Ambeed
Warning

A325037-100MG A325037-250MG A325037-5G
77032-792EA 45.62 USD
77032-792 77032-794 77668-458
4-Bromobenzo[d]isoxazole 97%
4-Bromobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₄BrNO
MW: 198.0168 g/mol
MDL Number: MFCD09027766
CAS Number: 1126848-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR