8-Chlorodibenzo[b,f]thiepin-10(11H)-one 95+%

Supplier: Ambeed
Warning

A324689-100G A324689-25G A324689-5G
76989-188EA 149 USD
76989-188 76989-190 76989-192
8-Chlorodibenzo[b,f]thiepin-10(11H)-one 95+%
8-Chlorodibenzo[b,f]thiepin-10(11H)-one

Precaución: for Research Use Only

Formula: C₁₄H₉ClOS
MDL Number: MFCD01411917
CAS Number: 1469-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR