Ethyl 6-bromoquinoline-3-carboxylate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A324241-100MG A324241-250MG A324241-1G
76989-178EA 14 USD
76989-178 76989-180 77683-965
Ethyl 6-bromoquinoline-3-carboxylate 95%, Ambeed.Inc
Ethyl 6-bromoquinoline-3-carboxylate

Precaución: For research use only.

Formula: C₁₂H₁₀BrNO₂
MDL Number: MFCD11877875
CAS Number: 481054-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR