2-Methylbenzo[b]thiophene-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methylbenzo[b]thiophene-3-carbaldehyde

A323767-1G A323767-5G
77015-310EA 125.19 USD
77015-310 77015-312
2-Methylbenzo[b]thiophene-3-carboxaldehyde 95%
2-Methylbenzo[b]thiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈OS
MW: 176.235 g/mol
MDL Number: MFCD01651753
CAS Number: 30446-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR