tert-Butyl-2-chloro-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-chloro-4-fluorobenzoate

A323737-250MG A323737-5G
76989-170EA 5 USD
76989-170 77890-647
tert-Butyl-2-chloro-4-fluorobenzoate 98%
tert-Butyl 2-chloro-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₂
MDL Number: MFCD09800898
CAS Number: 911314-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR