Fmoc-D-alaninol 98%

Supplier: Ambeed
Warning

A323217-250MG A323217-1G A323217-25G A323217-5G
77002-888EA 8 USD
77002-888 77601-450 77881-904 77881-905
Fmoc-D-alaninol 98%
Fmoc-D-alaninol

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₃
MDL Number: MFCD00270234
CAS Number: 202751-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR