Methyl-4-methoxy-2-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-methoxy-2-nitrobenzoate, 4-Methoxy-2-nitrobenzoic acid methyl ester
A322999-10G
A322999-1G
A322999-250MG
A322999-25G
A322999-5G
77015-278EA
227.04
USD
77015-278
77015-280
77015-282
77015-284
77015-286
Methyl-4-methoxy-2-nitrobenzoate 97%
Methyl-4-methoxy-2-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉NO₅ MW: 211.17 g/mol |
MDL Number:
MFCD06658482 CAS Number: 181871-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...