7-Chloroisoquinolin-3(2H)-one 97%

Supplier: Ambeed
Warning

A322954-100MG A322954-1G A322954-250MG
77015-272EA 65 USD
77015-272 77015-274 77015-276
7-Chloroisoquinolin-3(2H)-one 97%
7-Chloroisoquinolin-3(2H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD19704528
CAS Number: 1175272-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR