3-Chloro-4-(2'-fluorobenzyloxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-4-((2-fluorobenzyl)oxy)phenyl)boronic acid

A322455-1G A322455-250MG
77015-268EA 16 USD
77015-268 77015-270
3-Chloro-4-(2'-fluorobenzyloxy)phenylboronic acid 98%
3-Chloro-4-(2'-fluorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BClFO₃
MW: 280.4900 g/mol
MDL Number: MFCD06798082
CAS Number: 870777-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR