4-Chloro-6-(1h-imidazol-1-yl)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-(1H-imidazol-1-yl)pyrimidine

A322411-250MG A322411-5G
77015-264EA 35.01 USD
77015-264 77015-266
4-Chloro-6-(1h-imidazol-1-yl)pyrimidine 97%
4-Chloro-6-(1h-imidazol-1-yl)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅ClN₄
MW: 180.594 g/mol
MDL Number: MFCD01631197
CAS Number: 114834-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR