4-Amino-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A321813-10G A321813-1G A321813-250MG A321813-25G A321813-5G
77002-878EA 43 USD
77002-878 77002-880 77002-882 77002-884 77002-886
4-Amino-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Amino-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD01686848
CAS Number: 1500-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR