(±)-3-Butene-1,2-diol 98+% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
But-3-ene-1,2-diol, 3,4-Dihydroxy-1-butene, (R,S)-1-Butene-3,4-diol, (±)-1-Butene-3,4-diol, (R,S)-3-Butene-1,2-diol
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A321732-100G
A321732-25G
A321732-5G
76989-162EA
83.82
USD
76989-162
76989-164
76989-166
(±)-3-Butene-1,2-diol 98+% stabilized
(±)-3-Butene-1,2-diol
Precaución: for Research Use Only
Formula:
C₄H₈O₂ MW: 88.11 g/mol |
MDL Number:
MFCD00239410 CAS Number: 497-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...