4-(Difluoromethyl)piperidine hydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A321646-1G A321646-100MG A321646-250MG A321646-5G
76989-160EA 166 USD
76989-160 77683-958 77885-854 77885-855
4-(Difluoromethyl)piperidine hydrochloride 97%, Ambeed.Inc
4-(Difluoromethyl)piperidine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₁F₂N·ClH
MDL Number: MFCD18711662
CAS Number: 1283717-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR