Indoline-5,6-diol hydrobromide 97%

Supplier: Ambeed
Warning

A321539-1G A321539-250MG
77002-870EA 45.62 USD
77002-870 77672-650
Indoline-5,6-diol hydrobromide 97%
Indoline-5,6-diol hydrobromide

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO₂
MW: 232.08 g/mol
MDL Number: MFCD07368401
CAS Number: 138937-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR