2-(Hydroxymethyl)-N-methylbenzamide 95+%

Supplier: Ambeed
Warning

A321523-1G A321523-250MG A321523-5G
77015-256EA 65 USD
77015-256 77015-258 77015-260
2-(Hydroxymethyl)-N-methylbenzamide 95+%
2-(Hydroxymethyl)-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD00157782
CAS Number: 39976-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR