(±)-1-Benzyl-3-hydroxypiperidine hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3-piperidinol hydrochloride

A321487-5G A321487-100G A321487-1G A321487-25G
77002-860EA 15 USD
77002-860 77881-901 77881-902 77881-903
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride 98+%
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClNO
MW: 227.7304 g/mol
MDL Number: MFCD00044200
CAS Number: 105973-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR