5-Bromo-2,4,6-trichloropyrimidine 95%

Supplier: Ambeed
Warning

A321427-100MG A321427-10G A321427-1G A321427-250MG A321427-5G
76989-150EA 10 USD
76989-150 76989-152 76989-154 76989-156 76989-158
5-Bromo-2,4,6-trichloropyrimidine 95%
5-Bromo-2,4,6-trichloropyrimidine

Precaución: for Research Use Only

Formula: C₄BrCl₃N₂
MDL Number: MFCD11226676
CAS Number: 63931-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR