11,12-Dihydro-11-phenylindolo[2,3-a]carbazole 95%

Supplier: Ambeed
Warning

Synonyms: 11-Phenyl-11,12-dihydroindolo[2,3-a]carbazole, 11-Phenylindolo[2,3-a]carbazole

A321259-100MG A321259-1G A321259-250MG A321259-5G
76989-142EA 6.37 USD
76989-142 76989-144 76989-146 76989-148
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole 95%
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂
MW: 332.41 g/mol
MDL Number: MFCD22581305
CAS Number: 1024598-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR