(3-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A321197-1G A321197-250MG A321197-5G
77015-250EA 37.14 USD
77015-250 77015-252 77015-254
(3-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid 95%
(3-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₁BO₃
MW: 284.16 g/mol
MDL Number: MFCD08276820
CAS Number: 1072951-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR