Fmoc-4-amino-D-phenylalanine 98%

Supplier: Ambeed
Warning

A321006-1G A321006-250MG A321006-5G
76989-136EA 17 USD
76989-136 76989-138 76989-140
Fmoc-4-amino-D-phenylalanine 98%
Fmoc-4-amino-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₂O₄
MDL Number: MFCD02682658
CAS Number: 324017-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR