2-Bromo-6-chloropyrazine 98%

Supplier: Ambeed
Warning

A320993-250MG A320993-1G A320993-5G
77032-770EA 39 USD
77032-770 77593-900 77668-430
2-Bromo-6-chloropyrazine 98%
2-Bromo-6-chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
MDL Number: MFCD08752648
CAS Number: 916791-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR