3-Thiophenecarbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile

A320488-100G A320488-10G A320488-25G A320488-5G
76989-118EA 166.57 USD
76989-118 76989-120 76989-122 76989-124
3-Thiophenecarbonitrile 95%
3-Thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₃NS
MW: 109.15 g/mol
MDL Number: MFCD00151852
CAS Number: 1641-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR