(R)-(+)-4-Methoxy-α-methylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(4-Methoxyphenyl)ethan-1-amine, (R)-(+)-1-(4-Methoxyphenyl)ethylamine

A320158-1G A320158-250MG A320158-25G A320158-10G
77002-842EA 11.67 USD
77002-842 77002-844 77002-846 77593-894
(R)-(+)-4-Methoxy-α-methylbenzylamine 98%
(R)-(+)-4-Methoxy-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00671659
CAS Number: 22038-86-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR