O-Benzyl-L-threonine benzyl ester oxalate 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-Benzyl 2-amino-3-(benzyloxy)butanoate oxalate, H-Thr(Bzl)-OBzl·oxalate (1:1)

A320079-1G A320079-250MG
77002-838EA 11 USD
77002-838 77002-840
O-Benzyl-L-threonine benzyl ester oxalate 95%
O-Benzyl-L-threonine benzyl ester oxalate

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₇
MDL Number: MFCD02259508
CAS Number: 15260-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR