(4-(Trifluoromethyl)pyrimidin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A320013-100MG A320013-1G A320013-250MG
76989-106EA 57.29 USD
76989-106 76989-108 76989-110
(4-(Trifluoromethyl)pyrimidin-2-yl)methanol 97%
(4-(Trifluoromethyl)pyrimidin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂O
MW: 178.112 g/mol
MDL Number: MFCD16987898
CAS Number: 1240594-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR