3-Methyl-4-(trifluoromethyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-(trifluoromethyl)benzeneboronic acid, (3-Methyl-4-(trifluoromethyl)phenyl)boronic acid

A319925-100MG A319925-1G A319925-250MG
77032-764EA 84.88 USD
77032-764 77032-766 77032-768
3-Methyl-4-(trifluoromethyl)phenylboronic acid 95%
3-Methyl-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
MW: 203.954 g/mol
MDL Number: MFCD08706260
CAS Number: 864759-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR