6-Chloro-4-iodopyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
A319732-100MG
A319732-1G
A319732-250MG
A319732-5G
77032-756EA
9
USD
77032-756
77872-450
77872-451
77872-452
6-Chloro-4-iodopyridin-3-amine 97%
6-Chloro-4-iodopyridin-3-amine
Precaución: for Research Use Only
Formula:
C₅H₄ClIN₂ |
MDL Number:
MFCD11520341 CAS Number: 351227-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...