2-Propylpiperidin-4-one hydrochloride 97%

Supplier: Ambeed
Warning

A319615-100MG A319615-1G A319615-250MG
77032-748EA 53.05 USD
77032-748 77032-750 77032-752
2-Propylpiperidin-4-one hydrochloride 97%
2-Propylpiperidin-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO
MW: 177.67 g/mol
MDL Number: MFCD11110451
CAS Number: 1245645-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR