5-Chloro-4-iodo-2-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A319357-1G A319357-250MG
76989-098EA 472.11 USD
76989-098 76989-100
5-Chloro-4-iodo-2-(trifluoromethyl)pyridine 95%
5-Chloro-4-iodo-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₃IN
MW: 307.44 g/mol
MDL Number: MFCD18255894
CAS Number: 823221-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR