2-Chloro-5-methylnicotinaldehyde 98%

Supplier: Ambeed
Warning

A319321-100MG A319321-1G A319321-250MG A319321-5G
76989-090EA 24 USD
76989-090 76989-092 76989-094 76989-096
2-Chloro-5-methylnicotinaldehyde 98%
2-Chloro-5-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MDL Number: MFCD07375370
CAS Number: 92444-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR