Methyl 2-(5-bromo-2-nitrophenyl)acetate 95%

Supplier: Ambeed
Danger

A319281-1G A319281-250MG
77015-246EA 124.13 USD
77015-246 77015-248
Methyl 2-(5-bromo-2-nitrophenyl)acetate 95%
Methyl 2-(5-bromo-2-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
MDL Number: MFCD11977366
CAS Number: 189748-25-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR