4-Hydroxy-5-methyl-3-furanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-5-methylfuran-3(2H)-one

A319231-100G A319231-25G A319231-5G
77002-812EA 183.54 USD
77002-812 77002-814 77002-816
4-Hydroxy-5-methyl-3-furanone 98%
4-Hydroxy-5-methyl-3-furanone

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.1 g/mol
MDL Number: MFCD02752619
CAS Number: 19322-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR