tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-oxo-2,7-diazaspiro[4.5]decane-7-carboxylate

A318630-100MG A318630-1G A318630-250MG
77032-726EA 42.44 USD
77032-726 77032-728 77032-730
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
MDL Number: MFCD11227060
CAS Number: 923009-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR