7-Bromobenzo[d]oxazol-2-amine 98%
Supplier: Ambeed
|
Warning
|
A318452-100MG
A318452-1G
A318452-250MG
76989-078EA
101.85
USD
76989-078
76989-080
76989-082
7-Bromobenzo[d]oxazol-2-amine 98%
7-Bromobenzo[d]oxazol-2-amine
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂O MW: 213.03 g/mol |
MDL Number:
MFCD19375775 CAS Number: 1211527-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...