2,2-Dimethyl-3-oxopentanal 98%

Supplier: Ambeed
Warning

A318097-5G A318097-1G A318097-250MG
76989-076EA 123 USD
76989-076 77890-635 77890-636
2,2-Dimethyl-3-oxopentanal 98%
2,2-Dimethyl-3-oxopentanal

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MDL Number: MFCD11846616
CAS Number: 106921-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR