2-Chloro-1,3-bis(dimethylamino)trimethiniumhexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: (Z)-N-[2-Chloro-3-(dimethylamino)allylidene]-N-methylmethanaminium Hexafluorophosphate

A317772-10G A317772-500G A317772-25G
77015-232EA 7.43 USD
77015-232 77593-886 77683-945
2-Chloro-1,3-bis(dimethylamino)trimethiniumhexafluorophosphate 98%
2-Chloro-1,3-bis(dimethylamino)trimethiniumhexafluorophosphate

Precaución: For research use only.

Formula: C₇H₁₄ClF₆N₂P
MW: 306.62 g/mol
MDL Number: MFCD08458255
CAS Number: 291756-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR