2-Aminobenzo[b]thiophene-3-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A317768-100MG
A317768-1G
A317768-250MG
77032-712EA
37
USD
77032-712
77032-714
77032-716
2-Aminobenzo[b]thiophene-3-carbonitrile 95%
2-Aminobenzo[b]thiophene-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₆N₂S |
MDL Number:
MFCD00464272 CAS Number: 18774-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...