5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde 98+%

Supplier: Ambeed
Warning

A317566-1G A317566-250MG
76989-070EA 360.71 USD
76989-070 76989-072
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde 98+%
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O
MW: 181.579 g/mol
MDL Number: MFCD22551530
CAS Number: 1256162-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR