2-Chloro-5-methylpyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-hydroxy-5-methylpyridine, 2-Chloro-3-hydroxy-5-picoline

A317478-100MG A317478-250MG
77002-802EA 79.57 USD
77002-802 77002-804
2-Chloro-5-methylpyridin-3-ol 98%
2-Chloro-5-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD09839272
CAS Number: 910649-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR